Learn More
Sino Biological Anti-Vitamin D Receptor/VDR Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 101023T08100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the human VDR, and purified by antigen affinity chromatography.
Catalog No. 50-161-2697