Learn More
Sino Biological Anti-XAB1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 101025T36100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human GPN1 / XAB1, and purified by antigen affinity chromatography.
Catalog No. 50-161-2703