Learn More
Sino Biological Recombinant Human CD34 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10103H08H50
Encompass_Preferred
A DNA sequence encoding the extracellular domain of human CD34 precursor (NP_001020280.1) (Met 1-Thr 290) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-4004