Learn More
Sino Biological Anti-DNA-PKcs/PRKDC Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 101038T10100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human PRKDC, and purified by antigen affinity chromatography.
Catalog No. 50-161-2724