Learn More
Sino Biological Recombinant Human ST2/IL-1 RL1 Protein (hFc Tag), HPLC-verified
Supplier:  Sino Biological 10105H02H100
Encompass_Preferred
A DNA sequence encoding the human IL1RL1 isoform 2 (NP_003847.2) (Met 1-Phe 328) was fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-4005