Learn More
Sino Biological Anti-ST2/IL-1 RL1 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10105MM12100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IL1RL1 / IL‑1 R4 (rh IL1RL1 / IL‑1 R4; Catalog#10105-H08H; NP_003847.2; Met1-Phe328). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-0980