Learn More
Sino Biological Anti-ST2/IL-1 RL1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10105RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human IL1RL1 / ST2 (rh IL1RL1 / ST2; Catalog#10105-H08H; Met1-Phe328; NP_003847.2). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-172-0985