Learn More
Sino Biological Anti-RAF1/c-Raf Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 101053T32100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human Raf-1, and purified by antigen affinity chromatography.
Catalog No. 50-172-0954