Learn More
Sino Biological Anti-RND3 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 101056T32100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human RND3, and purified by antigen affinity chromatography.
Catalog No. 50-172-0960