Learn More
Sino Biological Recombinant Anti-Carbonic Anhydrase IX/CA9 Antibody (PE), Rabbit Monoclonal
Supplier:  Sino Biological 10107R053P25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human Carbonic Anhydrase IX/CA9 (rh Carbonic Anhydrase IX/CA9; Catalog#10107-H08H; NP_001207.2; Met1-Asp414) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-172-1031