Learn More
Sino Biological Recombinant Human ACE2 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10108H08H20
Encompass_Preferred
A DNA sequence encoding the human ACE2 (NP_068576.1) (Met1-Ser740) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-4021