Learn More
Sino Biological Anti-ACE2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10108RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human ACE2 / Angiotensin-Converting Enzyme 2 (rh ACE2 / Angiotensin-Converting Enzyme 2; NP_068576.1; 1M-740S). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-172-1062