Learn More
Sino Biological Recombinant Human CD155/PVR Protein (ECD, hFc Tag), HPLC-verified
Catalog No. 501614022
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met1-Asn343) of human CD155 (P15151-1) was expressed with the C-terminal fused Fc region of human IgG1.
Catalog No. 50-161-4022 Supplier Sino Biological Supplier No. 10109H02H100
Add to Cart
Add to Cart
A DNA sequence encoding the extracellular domain (Met1-Asn343) of human CD155 (P15151-1) was expressed with the C-terminal fused Fc region of human IgG1.