Learn More
Sino Biological Recombinant Human CD155/PVR Protein (ECD, His Tag), HPLC-verified
Supplier:  Sino Biological 10109H08H100
Encompass_Preferred
A DNA sequence encoding the human CD155 isoform Alpha (P15151-1) extracellular domain (Met1-Asn343) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-4025