Learn More
Sino Biological Anti-CD155/PVR Antibody (FITC), Mouse Monoclonal
Supplier:  Sino Biological 10109MM09F25
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD155 / PVR (rh CD155 / PVR; Catalog#10109-H08H; P15151-1; Met 1-Asn 343) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-172-1077