Learn More
Sino Biological Recombinant Anti-CD155/PVR Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10109R114100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD155 extracellular domain (rh CD155; Catalog#10109-H08H; Met 1-Asn 343; P15151-1).
Catalog No. 50-172-1082