Learn More
Sino Biological Anti-CD155/PVR Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10109RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CD155 isoform a (rh CD155; Catalog#10109-H08H; P15151-1; Met 1-Asn 343). CD155 specific IgG was purified by human CD155 affinity chromatography.
Catalog No. 50-172-1087