Learn More
Abcam 10,11-Dehydrocurvularin, antibiotic and antimitotic agent, 5MG
Catalog No. 502705477
Encompass_Preferred
MW 290.31 Da, Purity >95%. Antibiotic. Antifungal. Nematicidal. Phytotoxic. TGF-beta signaling inhibitor. Anticancer compound. Strong activator of the heat shock response. Inhibits the appressorium formation of M. grisea.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5477 Supplier Abcam Supplier No. AB1448555MG
MW 290.31 Da, Purity >95%. Antibiotic. Antifungal. Nematicidal. Phytotoxic. TGF-beta signaling inhibitor. Anticancer compound. Strong activator of the heat shock response. Inhibits the appressorium formation of M. grisea.
The product is subject to the following: Abcam Restricted Use Statement