Learn More
Sino Biological Recombinant Human IL1R2 Protein (ECD, His Tag), HPLC-verified
Supplier:  Sino Biological 10111H08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Glu 343) of human IL1R2 (NP_004624.1) was expressed, with a fused C-terminal polyhistidine tag.
Catalog No. 50-161-4032