Learn More
Sino Biological Recombinant Anti-IL1R2 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10111R020100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human IL1R2 (rh IL1R2; Catalog#10111-H08H; Met 1-Glu 343; NP_004624.1).
Catalog No. 50-172-1139