Learn More
Sino Biological Anti-IL1R2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10111RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human IL1R2 (rh IL1R2; Catalog#10111-H08H; Met 1-Glu 343; NP_004624.1). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-172-1144