Learn More
Sino Biological Recombinant Human VCAM-1/CD106 Protein (ECD, hFc Tag), HPLC-verified
Supplier:  Sino Biological 10113H02H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Pro 697) of human VCAM1 (NP_001069.1) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-4036