Learn More
Sino Biological Recombinant Human DUSP3 Protein (His & GST Tag)
Supplier:  Sino Biological 10114H20B100
Encompass_Preferred
A DNA sequence encoding the human VHR (P51452) (Met 1-Pro 185) was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus.
Catalog No. 50-161-4042