Learn More
Sino Biological Anti-DUSP3 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10114T52100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human DUSP3/VHR (rh DUSP3/VHR; Catalog#10114-HNCB; P51452; Met1-Pro185). DUSP3/VHR specific IgG was purified by Human DUSP3/VHR affinity chromatography.
Catalog No. 50-172-1193