Learn More
Sino Biological Anti-YAP1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 101146T10100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human YAP1, and purified by antigen affinity chromatography.
Catalog No. 50-172-1187