Learn More
Sino Biological Recombinant Human CD146/MCAM Protein (hFc Tag)
Supplier:  Sino Biological 10115H02H50
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Gly 559) of human CD146 precursor (NP_006491.2) was expressed with the C-terminal fused Fc region of human IgG1.
Catalog No. 50-161-4047