Learn More
Sino Biological Recombinant Anti-CD146/MCAM Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10115R012100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD146 / MCAM extracellular domain (rh CD146; Catalog#10115-H08H; NP_006491.2; Met 1-Gly 559).
Catalog No. 50-172-1236