Learn More
Sino Biological Recombinant Human IL-18 Protein (pro form, GST Tag)
Supplier:  Sino Biological 10119H09E50
Encompass_Preferred
A DNA sequence encoding the pro form of human IL18 (Q14116) (Met1-Asp193) was fused with the GST tag at the N-terminus.
Catalog No. 50-161-4055