Learn More
Sino Biological Recombinant Human IL-18 Protein, HPLC-verified
Supplier:  Sino Biological 10119HNCE100
Encompass_Preferred
A DNA sequence encoding the human IL18 (NP_001553.1) (Tyr37-Asp193) was expressed.
Catalog No. 50-161-4056