Learn More
Sino Biological Anti-IL-18 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10119MM15100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IL18 / IL-18 / Interleukin 18 (rh IL18 / IL-18 / Interleukin 18; Catalog#10119-H09E; Q14116; Met1-Asp193). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-1322