Learn More
Sino Biological Anti-IL-18 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10119T52100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human IL-18 (rh IL-18; Catalog#10119-H09E; Q14116; Met1-Asp193). IL-18 specific IgG was purified by Human IL-18 affinity chromatography.
Catalog No. 50-172-1331