Learn More
Cayman Chemical 1012 ZConjUGated LInole 25mg
Supplier: Cayman Chemical 2458425 MG
A conjugated linoleic acid methyl ester; has been used as a standard in the quantification of 10(E),12(Z)-conjugated linoleic acid from L. plantarum culture samples