Learn More
Sino Biological Anti-JAML/AMICA Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10120RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human JAML (rh JAML; Catalog#10120-H08H; NP_001091996.1; Met 1-Leu 275). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-172-1366