Learn More
Sino Biological Anti-REA/PHB2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 101202T34100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human PHB2, and purified by antigen affinity chromatography.
Catalog No. 50-172-1340