Learn More
Sino Biological Anti-IL1RAP/IL-1RAcP Antibody (PE), Mouse Monoclonal
Supplier:  Sino Biological 10121MM05P25
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IL-1RAcP / IL-1R3 (rh IL-1RAcP / IL-1R3; Catalog#10121-H08H; NP_002173.1; Met 1-Glu 359) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-172-1395