Learn More
Sino Biological Anti-Sin3a Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 101214T08100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Mouse Sin3a, and purified by antigen affinity chromatography.
Catalog No. 50-172-1375