Learn More
Sino Biological Recombinant Human IL-1RA/IL-1RN Protein (hFc Tag)
Supplier:  Sino Biological 10123H01H50
Encompass_Preferred
A DNA sequence encoding the mature form of human IL1RA (NP_776214.1) (Arg 26-Glu 177) was fused with the Fc region of human IgG1 at the N-terminus.
Catalog No. 50-161-4068