Learn More
Sino Biological Anti-IL-1RA/IL-1RN Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10123MM02100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IL-1RA (rh IL-1RA; Catalog#10123-HNAE; NP_776214.1; Arg26-Glu177). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-1438