Learn More
Sino Biological Alpha-Tubulin Antibody, Rabbit PAb, Antigen Affinity Purified
Supplier:  Sino Biological 101235T10100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human Alpha-Tubulin, and purified by antigen affinity chromatography.
Catalog No. 50-172-1423