Learn More
Sino Biological Recombinant Human IL-36 gamma/IL-1 F9 Protein (aa 18-169, His Tag)
Supplier:  Sino Biological 10124H07E120
Encompass_Preferred
A DNA sequence encoding the mature form of human IL1F9 (NP_062564) (Ser18-Asp169) was expressed with a polyhistide tag at the N-terminus.
Catalog No. 50-161-4072