Learn More
Sino Biological Recombinant Human IL-36 gamma/IL-1 F9 Protein (aa 18-169)
Supplier:  Sino Biological 10124HNCE120
Encompass_Preferred
A DNA sequence encoding the human IL1F9 (NP_062564) (Ser18-Asp169) was expressed and purified, with two additional amino acids (Gly & Pro) at the N-terminus.
Catalog No. 50-161-4076