Learn More
Sino Biological Anti-IL-36 gamma/IL-1 F9 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10124T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human IL36G (rh IL36G; Catalog#10124-H07E1; NP_062564; Ser18-Asp169). IL36G specific IgG was purified by Human IL36G affinity chromatography.
Catalog No. 50-172-1495