Learn More
Sino Biological Anti-Cofilin 2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 101244T46100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human cofilin 2/CFL2, and purified by antigen affinity chromatography.
Catalog No. 50-172-1472