Learn More
Sino Biological Anti-Islet-2/ISL2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 101249T34100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human ISL2, and purified by antigen affinity chromatography.
Catalog No. 50-172-1484