Learn More
Sino Biological Recombinant Human IL1R1 Protein
Supplier:  Sino Biological 10126HCCH100
Encompass_Preferred
A DNA sequence encoding the human IL1R1 (NP_000868.1) (Met1-Thr332) was expressed with five amino acids (DDDDK) at the C-terminus was expressed and purified.
Catalog No. 50-161-4083