Learn More
Sino Biological Anti-IL1R1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10126RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human IL1R1 (rhIL1R1; Catalog#10126-H08H; NP_000868.1; Met 1-Thr 332). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-172-1542