Learn More
Sino Biological Anti-RBM15 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 101269T36100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human RBM15 / OTT1, and purified by antigen affinity chromatography.
Catalog No. 50-172-1530