Learn More
Sino Biological Anti-beta-III Tubulin/TUBB3 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 101288T34100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human TUBB3 / beta III Tubulin, and purified by antigen affinity chromatography.
Catalog No. 50-172-1566