Learn More
Sino Biological Anti-PCSK1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10129RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human PCSK1 (rh PCSK1; Catalog#10129-H08H; NP_000430.3; Met 1-Arg 617). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-172-1600