Learn More
Sino Biological Anti-S-tag Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 101290T38500
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the S-tag sequence (KETAAAKFERQHMDS), and purified by antigen affinity chromatography.
Catalog No. 50-172-1587